missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hexafluorophosphate, 99%, Thermo Scientific™

Product Code. 10520414 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10520414 100 g Glass bottle 100g
10264850 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10520414 Supplier Thermo Scientific Acros Supplier No. 138341000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-32-7
Molecular Formula C4H12F6NP
Molecular Weight (g/mol) 219.111
MDL Number MFCD00012012
InChI Key ZFKULDQWLXAKIM-UHFFFAOYSA-N
Synonym tetramethylammonium hexafluorophosphate, ammonium, tetramethyl-, hexafluorophosphate 1-, tetramethylazanium hexafluorophosphate, c4h12n.f6p, acmc-209lpp, hexafluoro-$l^ 5-phosphanuide; tetramethylammonium ion, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-, methanaminium, n,n,n-trimethyl-, hexafluorophosphate 1-9ci
PubChem CID 11209
IUPAC Name tetramethylazanium;hexafluorophosphate
SMILES C[N+](C)(C)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point >300.0°C
Color White
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)4NPF6
Refractive Index 1.4820 to 1.4850
Quantity 100 g
Formula Weight 219.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tetramethylammonium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • Tetramethylammonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.