missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide, 10% in Water

Product Code. 10167850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10167850 100 g Plastic bottle 100g
10712032 500 g Plastic bottle 500g
10742784 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10167850 Supplier Thermo Scientific Acros Supplier No. 420511000
  • ONLINE EXCLUSIVE PRICE
    31.75 €
    30.80 € / 100g
    Save 0.95 €
    2% Off
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-59-2 | C4H13NO | 91.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium hydroxide
Name Note 10% in Water
CAS 75-59-2, 7732-18-5
Color Colorless
Density 1.0000g/mL
Boiling Point 100.0°C
Packaging Plastic bottle
Molecular Formula C4H13NO
Linear Formula (CH3)4NOH
MDL Number MFCD00008280
Quantity 100 g
Beilstein 04, 50
Fieser 11,514
Merck Index 15, 9371
Specific Gravity 1
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
Solubility Information Solubility in water: soluble.
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.[OH-]
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.15
PubChem CID 60966
Formula Weight 91.15
Percent Purity 9.5 to 10.5%
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium hydroxide, 10% in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Tetramethylammoniumhydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.