missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Hydroxide, 10% in Water
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Chemical Name or Material | Tetramethylammonium hydroxide |
| Name Note | 10% in Water |
| CAS | 75-59-2, 7732-18-5 |
| Color | Colorless |
| Density | 1.0000g/mL |
| Boiling Point | 100.0°C |
| Packaging | Plastic bottle |
| Molecular Formula | C4H13NO |
| Linear Formula | (CH3)4NOH |
| MDL Number | MFCD00008280 |
| Show More |
Safety and Handling
Product Identifier
- Tetramethylammonium hydroxide, 10% in water
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Tetramethylammoniumhydroxide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction