missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide pentahydrate, 97%

Product Code. 10134550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10134550 100 g Glass bottle 100g
10606442 25 g Glass bottle 25g
10609443 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10134550 Supplier Thermo Scientific Acros Supplier No. 207521000
  • ONLINE EXCLUSIVE PRICE
    158.00 €
    148.00 € / 100g
    Save 10.00 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10424-65-4 | C4H23NO6 | 181.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10424-65-4
Molecular Formula C4H23NO6
Molecular Weight (g/mol) 181.23
MDL Number MFCD00149566
InChI Key MYXKPFMQWULLOH-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
PubChem CID 82620
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C

Specifications

Melting Point 62.0°C to 71.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)4NOH·5H2O
Quantity 100 g
Beilstein 04, 50
Solubility Information Solubility in water: soluble.
Formula Weight 181.23
Percent Purity 97%
Physical Form Hygroscopic Crystals
Chemical Name or Material Tetramethylammonium hydroxide pentahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium hydroxide pentahydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P264-Wash thoroughly after handling.
Supplemental information
  • MIXTURE LIST-Contain: Methanaminium, N, N,N-trimethyl-, hydroxide, pentahydrate,Tetramethylammoniumhydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.