missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide pentahydrate, 98%

Product Code. 11342187
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342187 100 g 100g
11332187 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342187 Supplier Thermo Scientific Alfa Aesar Supplier No. L09658.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10424-65-4 | C4H23NO6 | 181.23 g/mol

Tetramethylammonium hydroxide pentahydrate is used in analytical chemistry as an ion pairing agent and a polarographic reagent. It finds application in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis and liquid chromatography. Furthermore, it is utilized in the preparation of iron oxide nanoparticles which are useful as magnetic resonance imaging contrast agents and tetramethylammonium dithiocarboxylates. It acts as a developer or an etchant in semiconductor and photoelectric industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium hydroxide pentahydrate is used in analytical chemistry as an ion pairing agent and a polarographic reagent. It finds application in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis and liquid chromatography. Furthermore, it is utilized in the preparation of iron oxide nanoparticles which are useful as magnetic resonance imaging contrast agents and tetramethylammonium dithiocarboxylates. It acts as a developer or an etchant in semiconductor and photoelectric industries.

Solubility
Soluble in water and methanol.

Notes
Air sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10424-65-4
Molecular Formula C4H23NO6
Molecular Weight (g/mol) 181.23
MDL Number MFCD00149566
InChI Key MYXKPFMQWULLOH-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
PubChem CID 82620
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C

Specifications

Melting Point 66°C to 67°C
Linear Formula (CH3)4NOH·5H2O
Quantity 100 g
UN Number UN3423
Beilstein 3714235
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,9224
Solubility Information Soluble in water and methanol.
Formula Weight 181.23 (91.15 Anhydrous)
Percent Purity 98%
Chemical Name or Material Tetramethylammonium hydroxide pentahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium hydroxide pentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tetramethylammoniumhydroxide pentahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.