missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide Pentahydrate, TRC

Product Code. 30416511
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30416511 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30416511

Brand: Toronto Research Chemicals TRCT07649525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium Hydroxide Pentahydrate
CAS 10424-65-4
Molecular Formula C4H23NO6
Quantity 25 g
Synonym Methanaminium','N,N,N-trimethyl-','hydroxide','hydrate (1:1:5) (ACI), Ammonium','tetramethyl-','hydroxide','pentahydrate (8CI), Methanaminium','N,N,N-trimethyl-','hydroxide','pentahydrate (9CI), Tetramethylammonium hydroxide pentahydrate
InChI Formula InChI=1S/C4H12N.6H2O/c1-5(2,3)4;;;;;;/h1-4H3;6*1H2/q+1;;;;;;/p-1
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
Molecular Weight (g/mol) 181.23
Formula Weight 181.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.