missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide Pentahydrate, TRC

Product Code. 30409917
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409917 Supplier Toronto Research Chemicals Supplier No. TRCT0764955G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium Hydroxide Pentahydrate
CAS 10424-65-4
Molecular Formula C4H23NO6
Quantity 5 g
Synonym Methanaminium','N,N,N-trimethyl-','hydroxide','hydrate (1:1:5) (ACI), Ammonium','tetramethyl-','hydroxide','pentahydrate (8CI), Methanaminium','N,N,N-trimethyl-','hydroxide','pentahydrate (9CI), Tetramethylammonium hydroxide pentahydrate
InChI Formula InChI=1S/C4H12N.6H2O/c1-5(2,3)4;;;;;;/h1-4H3;6*1H2/q+1;;;;;;/p-1
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
Molecular Weight (g/mol) 181.23
Formula Weight 181.1525
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.