missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium perchlorate, 98%

Product Code. 10782402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10782402 25 g 25g
11955451 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10782402 Supplier Thermo Scientific Acros Supplier No. 420560250
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    133.00 € / 25g
    Save 4.00 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 2537-36-2 | C4H12ClNO4 | 173.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2537-36-2
Molecular Formula C4H12ClNO4
Molecular Weight (g/mol) 173.59
MDL Number MFCD00043207
InChI Key ZCWKIFAQRXNZCH-UHFFFAOYSA-M
Synonym tetramethylammonium perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate, n,n,n-trimethylmethanaminium perchlorate, ammonium, tetramethyl-, perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate 1:1, c4h12n.clo4, acmc-20ajj6, tetramethylazanium perchlorate, tetramethylammonium perchlorate 25g, wln: 2k2&2&2 &wgw
PubChem CID 17337
IUPAC Name tetramethylazanium;perchlorate
SMILES C[N+](C)(C)C.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dry base)
Packaging Plastic bottle
Linear Formula (CH3)4NClO4
Quantity 25 g
Formula Weight 173.6
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetramethylammonium perchlorate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium perchlorate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.