missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium tetrafluoroborate, 97%

Product Code. 11498017
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498017 25 g 25g
11488017 5 g 5g
11408027 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498017 Supplier Thermo Scientific Alfa Aesar Supplier No. A17436.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 661-36-9 | C4H12BF4N | 160.95 g/mol

It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

Solubility
Solubility in hot water is almost transparent.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 661-36-9
Molecular Formula C4H12BF4N
Molecular Weight (g/mol) 160.95
MDL Number MFCD00011745
InChI Key XWFABLFRYCHILB-UHFFFAOYSA-N
Synonym tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate
PubChem CID 12621
IUPAC Name tetramethylazanium;tetrafluoroborate
SMILES C[N+](C)(C)C.F[B-](F)(F)F

Especificaciones

Melting Point 417°C to 419°C
Quantity 25 g
Beilstein 3715404
Sensitivity Hygroscopic
Formula Weight 160.95
Percent Purity 97%
Chemical Name or Material Tetramethylammonium tetrafluoroborate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.