missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Tetrafluoroborate, TRC

Product Code. 30457118
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457118 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30457118 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT889868250MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium Tetrafluoroborate
CAS 661-36-9
Molecular Formula C4H12BF4N
Quantity 250 mg
Synonym Methanaminium','N,N,N-trimethyl-','tetrafluoroborate(1-) (1:1) (ACI), Ammonium compounds','substituted','tetramethyl-','tetrafluoroborate (7CI), Ammonium','tetramethyl-','tetrafluoroborate(1-) (8CI), Borate(1-)','tetrafluoro-','N,N,N-trimethylmethanaminium (ZCI), Borate(1-)','tetrafluoro-','tetramethylammonium (8CI), Methanaminium','N,N,N-trimethyl-','tetrafluoroborate(1-) (9CI), Tetramethylammonium fluoroborate (6CI), [Me4N][BF4], Tetramethylammonium tetrafluoroborate, Tetramethylammonium tetrafluoroborate(1-)
InChI Formula InChI=1S/C4H12N.BF4/c1-5(2,3)4;2-1(3,4)5/h1-4H3;/q+1;-1
SMILES C[N+](C)(C)C.F[B-](F)(F)F
IUPAC Name tetramethylazanium;tetrafluoroborate
Molecular Weight (g/mol) 160.95
Formula Weight 161.0999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.