missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium triacetoxyborohydride, 90+%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Melting Point | 95.0°C to 98.0°C |
| Assay Percent Range | 90+% |
| Packaging | Glass bottle |
| Quantity | 50 g |
| Solubility Information | Solubility in water: .. Other solubilities: soluble in common organic solvents |
| Formula Weight | 263.1 |
| Percent Purity | 90+% |
| Chemical Name or Material | Tetramethylammonium triacetoxyborohydride |
Safety and Handling
Product Identifier
- Tetramethylammonium triacetoxyborohydride
- Danger
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction