missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylcyclobutane-1,3-dione, 99%

Product Code. 11487757
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11487757

Brand: Thermo Scientific Alfa Aesar A16297.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 933-52-8 | C8H12O2 | 140.182 g/mol

Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

Solubility
It is soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 933-52-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001331
InChI Key RGCDVHNITQEYPO-UHFFFAOYSA-N
Synonym tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
PubChem CID 13617
IUPAC Name 2,2,4,4-tetramethylcyclobutane-1,3-dione
SMILES CC1(C(=O)C(C1=O)(C)C)C

Specifications

Melting Point 112°C to 115°C
Boiling Point 120°C to 122°C (150 mmHg)
Quantity 50 g
Solubility Information It is soluble in chloroform.
Formula Weight 140.18
Percent Purity 99%
Chemical Name or Material Tetramethylcyclobutane-1,3-dione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.