missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylcyclopentadiene, mixed isomers, tech. 85%

Product Code. 11328887
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11328887

Brand: Thermo Scientific Alfa Aesar 038577.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4249-10-9 | C9H14 | 122.21 g/mol

Tetramethylcyclopentadiene is used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4249-10-9
Molecular Formula C9H14
Molecular Weight (g/mol) 122.21
MDL Number MFCD00145385
InChI Key VNPQQEYMXYCAEZ-UHFFFAOYSA-N
Synonym 1,2,3,4-tetramethylcyclopentadiene, tetramethylcyclopentadiene, 1,2,3,4-tetramethyl-1,3-cyclopentadiene, tetramethyl cyclopentadiene, tetramethylcyclopentadiene, mixed isomers, pubchem15072, acmc-1an6q, 1,3-cyclopentadiene, tetramethyl, 2,3,4,5-tetramethylcyclopenta-2,4-diene
PubChem CID 138163
IUPAC Name 1,2,3,4-tetramethylcyclopenta-1,3-diene
SMILES CC1=C(C(=C(C1)C)C)C

Specifications

Color Colorless
Density 0.808 g/mL
Boiling Point 55°C to 59°C (13 mmHg)
Flash Point 42°C (108°F)
Refractive Index 1.472
Quantity 5 g
UN Number UN3295
Sensitivity Air and moisture sensitive
Solubility Information Not miscible mix with mater.
Formula Weight 122.21
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tetramethylcyclopentadiene, Mixed isomers
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.