missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylene glutarimide, 99%, Thermo Scientific™

Product Code. 10541491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10541491 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10541491 Supplier Thermo Scientific Acros Supplier No. 255100250
Quantity Price
1 107.00 € / 25g
10 94.00 € / 25g

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

CAS 1075-89-4
Melting Point 153.0°C to 155.0°C
Color White
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H13NO2
MDL Number MFCD00023871
Quantity 25 g
Synonym 3,3-tetramethyleneglutarimide, 8-azaspiro 4.5 decane-7,9-dione, azaspirodecanedione, 1,1-cyclopentanediacetimide, 8-aza-spiro 4.5 decane-7,9-dione, buspirone impurity k, dsstox_cid_31405, dsstox_rid_97291, dsstox_gsid_57616
InChI Key YRTHJMQKDCXPAY-UHFFFAOYSA-N
SMILES C1CCC2(C1)CC(=O)NC(=O)C2
IUPAC Name 8-azaspiro[4.5]decane-7,9-dione
Molecular Weight (g/mol) 167.21
PubChem CID 136843
Formula Weight 167.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylene glutarimide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetramethylene glutarimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.