missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylene sulfone, 99+%

Product Code. 10225000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10225000 250 mL Glass bottle 250mL
10627943 1 L Glass bottle 1 L
11914561 2.5 L Glass bottle 2.5 L
15129183 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10225000 Supplier Thermo Scientific Acros Supplier No. 138432500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 126-33-0 | C4H8O2S | 120.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
PubChem CID 31347
ChEBI CHEBI:74794
IUPAC Name thiolane 1,1-dioxide
SMILES O=S1(=O)CCCC1

Specifications

Melting Point 27.4°C to 27.8°C
Color Yellow
Density 1.2600g/mL
Boiling Point 285.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4820 to 1.4850
Quantity 250 mL
Beilstein 17,I,5
Fieser 01,1144; 02,402; 05,651; 13,256
Merck Index 15,9085
Specific Gravity 1.26
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diluted mineral acids, soluble in acetone, benzene and toluene, freely soluble in alcohol
Viscosity 10.34 mPa.s (30°C)
Formula Weight 120.17
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material Tetramethylene sulfone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetramethylene sulfone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.