missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylene sulfone, 99+%

Product Code. 15129183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15129183 10 L Plastic drum 10 L
10225000 250 mL Glass bottle 250mL
10627943 1 L Glass bottle 1 L
11914561 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Artikelnummer. 15129183 Lieferant Thermo Scientific Acros Lieferanten-Nr. 138430100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

CAS: 126-33-0 | C4H8O2S | 120.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
PubChem CID 31347
ChEBI CHEBI:74794
IUPAC Name thiolane 1,1-dioxide
SMILES O=S1(=O)CCCC1

Spezifikation

Melting Point 27.4°C to 27.8°C
Color Yellow
Density 1.2600g/mL
Boiling Point 285.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Refractive Index 1.4820 to 1.4850
Quantity 10 L
Beilstein 17,I,5
Fieser 01,1144; 02,402; 05,651; 13,256
Merck Index 15,9085
Specific Gravity 1.26
Solubility Information Solubility in water: soluble. Other solubilities: soluble in diluted mineral acids, soluble in acetone, benzene and toluene, freely soluble in alcohol
Viscosity 10.34 mPa.s (30°C)
Formula Weight 120.17
Percent Purity 99+%
Physical Form Liquid After Melting
Chemical Name or Material Tetramethylene sulfone
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Tetramethylene sulfone
Signalwort
  • Danger||Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Sicherheitshinweis
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.