missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylethylene, 97%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Beschreibung
Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.
Solubility
Soluble in water (0.071 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.
Solubility
Soluble in water (0.071 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
Spezifikation
Spezifikation
| Melting Point | -75°C |
| Density | 0.705 |
| Boiling Point | 72°C to 73°C |
| Flash Point | −17°C (1°F) |
| Odor | Mild |
| Refractive Index | 1.412 |
| Quantity | 25 g |
| UN Number | UN3295 |
| Beilstein | 1361357 |
| Sensitivity | Air Sensitive |
| Mehr anzeigen |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur