missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylgermanium, 98%

Product Code. 11488756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488756 5 g 5g
11498756 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488756 Supplier Thermo Scientific Alfa Aesar Supplier No. A16931.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 865-52-1 | C4H12Ge | 132.77 g/mol

Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a Chemical Vapor Deposition (CVD) precursor to germanium oxide thin films.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-52-1
Molecular Formula C4H12Ge
Molecular Weight (g/mol) 132.77
MDL Number MFCD00014843
InChI Key ZRLCXMPFXYVHGS-UHFFFAOYSA-N
Synonym tetramethylgermanium, germanium, tetramethyl, germane, tetramethyl, tetramethyl germane, tetramethyl germanium, acmc-1bk8h, germanium tetramethyl, ch3 4ge, 4-04-00-04282 beilstein handbook reference
PubChem CID 70079
IUPAC Name tetramethylgermane
SMILES C[Ge](C)(C)C

Specifications

Melting Point -88°C
Density 0.975
Boiling Point 42°C to 44°C
Flash Point −37°C (−35°F)
Refractive Index 1.389
Quantity 5 g
UN Number UN1993
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 132.73
Percent Purity 98%
Chemical Name or Material Tetramethylgermanium
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetramethylgermanium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: Tetramethylgermanium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.