missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylpyrazine, TRC

Product Code. 30454539
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454539 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454539 Supplier Toronto Research Chemicals Supplier No. TRCT30464510G
 Restricted Item
Estimated Shipment: 03-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 2,3,5,6-Tetramethylpyrazine
CAS 1124-11-4
Molecular Formula C8 H12 N2
Quantity 10 g
Synonym Pyrazine','2,3,5,6-tetramethyl-, Pyrazine','tetramethyl- (6CI,7CI,8CI,9CI), 2,3,5,6-Tetramethylpyrazine, BS factor, Chuanxingzine, Ligustrazin, Ligustrazine, NSC 36080, NSC 46451, Tetramethylpyrazine, Tetrapyrazine
InChI Formula InChI=1S/C8H12N2/c1-5-6(2)10-8(4)7(3)9-5/h1-4H3
SMILES Cc1nc(C)c(C)nc1C
IUPAC Name 2,3,5,6-tetramethylpyrazine
Molecular Weight (g/mol) 136.19
Formula Weight 136.1
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.