missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsilane, 99%, AcroSeal™

Product Code. 10205004 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10205004 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10205004 Supplier Thermo Scientific Acros Supplier No. 427331000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75-76-3 | C4H12Si | 88.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 75-76-3
Melting Point -99°C
Color Colorless
Density 0.6400g/mL
Boiling Point 26°C to 28°C
Flash Point -27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C4H12Si
Linear Formula Si(CH3)4
Refractive Index 1.3585
MDL Number MFCD00008274
Quantity 100 mL
Merck Index 15, 9377
Specific Gravity 0.64
Synonym silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
Solubility Information Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents
InChI Key CZDYPVPMEAXLPK-UHFFFAOYSA-N
SMILES C[Si](C)(C)C
IUPAC Name tetramethylsilane
Molecular Weight (g/mol) 88.23
PubChem CID 6396
ChEBI CHEBI:85361
Formula Weight 88.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetramethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.