missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylthiuram disulfide, 97%

Product Code. 10563301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10563301 1 kg Plastic bottle 1kg
10113380 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10563301 Supplier Thermo Scientific Acros Supplier No. 138480010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-26-8 | C6H12N2S4 | 240.416 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-26-8
Molecular Formula C6H12N2S4
Molecular Weight (g/mol) 240.416
MDL Number MFCD00008325
InChI Key KUAZQDVKQLNFPE-UHFFFAOYSA-N
Synonym thiram, tetramethylthiuram disulfide, thiuram, tmtd, pomarsol, thirame, arasan, fernasan, nobecutan, rezifilm
PubChem CID 5455
ChEBI CHEBI:9495
IUPAC Name dimethylcarbamothioylsulfanyl N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)SSC(=S)N(C)C

Specifications

Melting Point >142°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Linear Formula [(CH3)2NCS2]2
Quantity 1 kg
Beilstein 04, 76
Merck Index 15, 9525
Formula Weight 240.41
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material Tetramethylthiuram disulfide, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylthiuram disulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.