missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylthiuram monosulfide, 97%

Product Code. 11433487
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433487 100 g 100g
11423487 25 g 25g
11443487 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433487 Supplier Thermo Scientific Alfa Aesar Supplier No. A17259.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97-74-5 | C6H12N2S3 | 208.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-74-5
Molecular Formula C6H12N2S3
Molecular Weight (g/mol) 208.356
MDL Number MFCD00014870
InChI Key REQPQFUJGGOFQL-UHFFFAOYSA-N
Synonym tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm
PubChem CID 7347
IUPAC Name dimethylcarbamothioyl N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)SC(=S)N(C)C

Specifications

Melting Point 106°C to 110°C
Density 1.37
Quantity 100 g
Beilstein 1775650
Formula Weight 208.37
Percent Purity 97%
Chemical Name or Material Tetramethylthiuram monosulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.