missing translation for 'onlineSavingsMsg'
Learn More

Tetramisole hydrochloride, 99.6%, MP Biomedicals™

Product Code. 11468251 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468251 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11468251 Supplier MP Biomedicals Supplier No. 0215211910

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 83-67-0
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.167
InChI Key YAPQBXQYLJRXSA-UHFFFAOYSA-N
Synonym theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
PubChem CID 5429
ChEBI CHEBI:28946
IUPAC Name 3,7-dimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)NC(=O)N2C

Specifications

Assay Percent Range 0.996
Quantity 10 g
Formula Weight 240.8
Physical Form Powder
Chemical Name or Material Tetramisole Hydrochloride
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium Hydroxide Pentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361+P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.