missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylboron sodium, 99.5%

Product Code. 10205430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205430 25 g Glass bottle 25g
10725631 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205430 Supplier Thermo Scientific Acros Supplier No. 138520250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Tetraphenylboron sodium, 99.5%, C24H20BNa, CAS Number-143-66-8, borate 1-, tetraphenyl-, sodium, dotite kalibor, kalibor, kalignost, kariporu k, sodium tetraphenylborate, sodium tetraphenylborate 1-, sodium tetraphenylboride, sodium tetraphenylboron, tetraphenylboron sodium, 100g | CAS: 143-66-8 | C24H20BNa | 342.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
PubChem CID 2723787
IUPAC Name sodium;tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Color Colorless
Loss on Drying 0.5 % max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99.4 % min. (Total base)
Packaging Glass bottle
Linear Formula (C6H5)4BNa
Quantity 25 g
Merck Index 15, 8818
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether
Formula Weight 342.21
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Tetraphenylboron sodium, p.a.
Show More Show Less
compliance-icons
Product Identifier
  • Tetraphenylboron sodium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.