missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylboron sodium, ACS reagent

Product Code. 10311221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311221 100 g Glass bottle 100g
10351171 25 g Glass bottle 25g
11905881 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10311221 Supplier Thermo Scientific Acros Supplier No. 424451000
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 143-66-8 | C24H20BNa | 342.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
PubChem CID 2723787
IUPAC Name sodium tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300.0°C
Color Colorless to White
Physical Form Crystals or Granular Powder
Quantity 100 g
Linear Formula (C6H5)4BNa
Merck Index 15,8818
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether
Formula Weight 342.21
Grade ACS Reagent
Infrared Spectrum Authentic
Loss on Drying 0.5 % max. (105°C)
Packaging Glass bottle
Chemical Name or Material Tetraphenylboron sodium
Show More Show Less
compliance-icons
Product Identifier
  • Tetraphenylboron sodium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.