missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylethylene, 98%

Product Code. 11322715
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322715 25 g 25g
11312715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322715 Supplier Thermo Scientific Alfa Aesar Supplier No. L00222.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 632-51-9 | C26H20 | 332.446 g/mol

Tetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylethylene is used in construction and in the manufacture of medical equipment, packaging, and electrical appliances. And it is used to produce other chemical like 1,1,2,2-Tetraphenyl-ethane.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-51-9
Molecular Formula C26H20
Molecular Weight (g/mol) 332.446
MDL Number MFCD00004764
InChI Key JLZUZNKTTIRERF-UHFFFAOYSA-N
Synonym tetraphenylethylene, tetraphenylethene, 1,1,2,2-tetraphenylethene, ethylene, tetraphenyl, 1,1,2,2-tetraphenylethylene, benzene, 1,1',1,1'-1,2-ethenediylidene tetrakis, tetraphenylethylen, 1,2,2-triphenylvinyl benzene, 1,2,2-tetraphenylethylene, 1,1',1,1'-1,2-ethenediylidene tetrakisbenzene
PubChem CID 69437
IUPAC Name 1,2,2-triphenylethenylbenzene
SMILES C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 222°C to 226°C
Boiling Point 420°C
Quantity 25 g
Beilstein 1913548
Solubility Information Insoluble in water.
Formula Weight 332.45
Percent Purity 98%
Chemical Name or Material Tetraphenylethylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.