missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylmethane, 96%

Product Code. 11417726
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417726 1 g 1g
11427726 5 g 5g
11437726 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417726 Supplier Thermo Scientific Alfa Aesar Supplier No. A14706.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 630-76-2 | C25H20 | 320.435 g/mol

Tetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylmethane is used to prepare a tetrapyridone tecton, which forms a diamondoid network with large internal chambers. It is also used in the preparation of 1-{4-[tris-(4-acetyl-phenyl)-methyl]-phenyl}-ethanone by Friedel-Crafts acylation reaction with acetyl chloride using aluminum chloride as catalyst.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-76-2
Molecular Formula C25H20
Molecular Weight (g/mol) 320.435
MDL Number MFCD00014428
InChI Key PEQHIRFAKIASBK-UHFFFAOYSA-N
Synonym tetraphenylmethane, methane, tetraphenyl, tetraphenyl methane, benzene, 1,1',1,1'-methanetetrayltetrakis, triphenylmethyl benzene, benzene, 1,1',1,1'-methanethetrayltetrakis, acmc-20ajiz, pubchem10677
PubChem CID 12424
IUPAC Name tritylbenzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 280°C to 285°C
Boiling Point 431°C
Quantity 1 g
Beilstein 1913557
Solubility Information Insoluble in water.
Formula Weight 320.44
Percent Purity 96%
Chemical Name or Material Tetraphenylmethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.