missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenyltin, Thermo Scientific™

Product Code. 15442947
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442947 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15442947 Supplier Thermo Scientific Alfa Aesar Supplier No. 071117.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 595-90-4
Melting Point 224°C to 227°C
Boiling Point >420°C
Flash Point >110°C (230°F)
Odor Characteristic
Molecular Formula C24H20Sn
MDL Number MFCD00003006
Quantity 100 g
UN Number UN3146
Synonym tetraphenyltin, stannane, tetraphenyl, tetraphenyl tin, ccris 6326, tin, tetraphenyl-,, stannane,tetraphenyl, acmc-209mem
Solubility Information Insoluble in water.
InChI Key CRHIAMBJMSSNNM-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name tetraphenylstannane
Molecular Weight (g/mol) 427.13
PubChem CID 61146
Formula Weight 427.11
Physical Form Crystalline
Chemical Name or Material Tetraphenyltin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.