missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium perruthenate

Product Code. 10758833 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10758833

Brand: Thermo Scientific Acros 378392500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114615-82-6 | C12H28NO4Ru | 351.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114615-82-6
Molecular Formula C12H28NO4Ru
Molecular Weight (g/mol) 351.43
MDL Number MFCD00074914
InChI Key HUCLFLGLPCVDMZ-UHFFFAOYSA-M
Synonym 1-propanaminium, n,n,n-tripropyl-, t-4-tetraoxoruthenate 1-
PubChem CID 127020979
IUPAC Name ruthenium;tetrapropylazanium;hydroxide;trihydrate
SMILES CCC[N+](CCC)(CCC)CCC.O.O.O.[OH-].[Ru]

Specifications

Melting Point 160.0°C
Color Black
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 9386
Formula Weight 351.43
Physical Form Powder
Chemical Name or Material Tetrapropylammonium perruthenate
compliance-icons compliance-icons
Product Identifier
  • Tetrapropylammonium perruthenate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.