missing translation for 'onlineSavingsMsg'
Learn More

Tetratriacontane, 98%

Product Code. 10319457 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10319457

Brand: Thermo Scientific Acros 432070010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 14167-59-0 | C34H70 | 478.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 14167-59-0
Melting Point 73.9°C to 74.9°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C34H70
MDL Number MFCD00009412
Quantity 1 g
Synonym n-tetratriacontane, unii-94ua0738yx, tetratriacontane, analytical standard, tetratriacontane, acmc-1bniv
InChI Key GWVDBZWVFGFBCN-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
IUPAC Name tetratriacontane
Molecular Weight (g/mol) 478.93
PubChem CID 26519
Formula Weight 478.93
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material Tetratriacontane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.