missing translation for 'onlineSavingsMsg'
Learn More

Tetrazole, 3 to 4 wt.% solution in acetonitrile, AcroSeal™

Product Code. 11309601 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
This item is not returnable. View return policy

Product Code. 11309601

Brand: Thermo Scientific Acros 432991000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 288-94-8 | CH2N4 | 70.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrazole
CAS 288-94-8, 75-05-8
Color Cream to Orange
Density 0.8000g/mL
Boiling Point 77°C
Flash Point 5°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula CH2N4
Quantity 100 mL
Specific Gravity 0.8
Synonym 1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
InChI Key KJUGUADJHNHALS-UHFFFAOYSA-N
SMILES C1=NNN=N1
IUPAC Name 2H-tetrazole
Molecular Weight (g/mol) 70.05
PubChem CID 67519
ChEBI CHEBI:33193
Formula Weight 70.05
Physical Form Crystalline Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrazole, 3 to 4 wt% solution in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.