missing translation for 'onlineSavingsMsg'
Learn More

Tetrazolium Blue Chloride (>90%), TRC

Product Code. 30450243
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450243 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450243 Supplier Toronto Research Chemicals Supplier No. TRCT3107001G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrazolium Blue Chloride
CAS 1871-22-3
Molecular Formula C40H32N8O2 . 2Cl
Quantity 1 g
Synonym 2,2'-(3,3'-Dimethoxy[1,1'-biphenyl]-4,4'-diyl)bis[3,5-diphenyl-2H-tetrazolium','','Dichloride, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylylene)bis[2,5-diphenyl-2H-tetrazolium','Dichloride, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylylene)bis[2,5-diphenyl-2H-tetrazolium chloride], 2,2',5,5'-Tetraphenyl-3,3'-(3,3'-dimethoxy-4,4'-diphenylene)ditetrazolium Chloride, 2,2'-(m,m'-Dimethoxy-p,p'-biphenylene)bis(3,5-diphenyltetrazolium Chloride), 3,3'-(3,3'-Dimethoxy-4,4'-biphenylene)bis[2,5-diphenyl-2H-tetrazolium Chloride], 3,3'-Dianisolebis[4,4'-(3,5-diphenyl)tetrazolium Chloride], BT, BT (dye), Blue Tetrazolium, Blue Tetrazolium Chloride, Ditetrazolium Chloride, NSC 27623, Tetrazolium Blue
InChI Formula InChI=1S/C40H32N8O2.2ClH/c1-49-37-27-31(23-25-35(37)47-43-39(29-15-7-3-8-16-29)41-45(47)33-19-11-5-12-20-33)32-24-26-36(38(28-32)50-2)48-44-40(30-17-9-4-10-18-30)42-46(48)34-21-13-6-14-22-34;;/h3-28H,1-2H3;2*1H/q+2;;/p-2
SMILES [Cl-].[Cl-].COc1cc(ccc1[n+]2nc(nn2c3ccccc3)c4ccccc4)c5ccc(c(OC)c5)[n+]6nc(nn6c7ccccc7)c8ccccc8
IUPAC Name 2-[4-[4-(3,5-diphenyltetrazol-2-ium-2-yl)-3-methoxyphenyl]-2-methoxyphenyl]-3,5-diphenyltetrazol-2-ium;dichloride
Molecular Weight (g/mol) 727.6
Formula Weight 726.2025
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.