missing translation for 'onlineSavingsMsg'
Learn More

Tetrazolium Red, MedChemExpress

Product Code. 16491168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491168 Supplier Medchem Express Supplier No. HYD0714
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tetrazolium Red(2,3,5-Triphenyltetrazolium chloride; TPTZ) is used to visualize dehydrogenase enzyme activity; initially the tetrazolium solution is colorless but changes to red when it comes into contact with hydrogen.

Tetrazolium Red(2,3,5-Triphenyltetrazolium chloride

Specifications

Chemical Name or Material Tetrazolium Red
CAS 298-96-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H15ClN4
Quantity 5 g
Synonym 2,3,5-Triphenyltetrazolium chloride TPTZ TTC
Solubility Information H2O : 50 mg/mL (149.34 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (49.79 mM; Need ultrasonic)
SMILES C1(C2=CC=CC=C2)=NN(C3=CC=CC=C3)[N+](C4=CC=CC=C4)=N1.[Cl-]
Molecular Weight (g/mol) 334.8
Formula Weight 334.8
Percent Purity 98.07%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.