missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ Alpha™ 10 Sealed-Border Wipers

Product Code. 15614987 Shop All Texwipe Products
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in.
9 x 9 in. (22.86 x 22.86 cm)
Product Type:
Dry Wipe
Pre-wetted Wipe
Saturation Solution:
70% IPA, 30% WFI
70% Isopropyl Alcohol
Dry Wiper
Unit Size:
Pack of 100
Pack of 250
Pack of 400
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Product Type Saturation Solution Unit Size
15614987 9 x 9 in. (22.86 x 22.86 cm) Pre-wetted Wipe 70% Isopropyl Alcohol Pack of 400
11585355 12 x 12 in. Dry Wipe Dry Wiper Pack of 100
15604987 12 x 12 in. Pre-wetted Wipe 70% IPA, 30% WFI Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15614987 Supplier Texwipe™ Supplier No. TX3285
  • ONLINE EXCLUSIVE PRICE
    1930.00 €
    946.78 € / Pack of 400
    Save 983.22 €
    50% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For ISO Class 3 to 4 applications

  • Vectra™ processed—hands-free, continuous manufacturing purification and inspection
  • Double-knit, no-run interlock construction of 100% continuous-filament polyester
  • Patented sealed-border wiper for lowest releasable fibers and particles
  • Optimum balance of wiping efficiency, abrasion resistance, and sorbency
  • Ideal wiper for use in multiple environments
  • Lot-to-lot traceability

Order Info

Available dry or sterile and presaturated with isopropanol. Basis weight: 130g/m2. Sterile wipers 19-130-2061 packed 50 wipers/ flatpack, 5 flatpacks /case; 19-165-653 packed 20 wipers per flexpack, 20 flexpacks/case.

Specifications

Product Type Pre-wetted Wipe
Length (English) 9 in.
Length (Metric) 22.9 cm
Width (English) 9 in.
Width (Metric) 22.9 cm
Material Polyester
Certifications/Compliance ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Edges Sealed
Dimensions (L x W) 9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application) Ideal wipes for use in multiple environments
Saturation Solution 70% Isopropyl Alcohol
Sterility Sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • wipers pre-wetted with 40% - 100% IPA
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.