missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Texwipe™ AlphaSorb™ 10 Sealed-Border Polyester Wipers
Shop All Texwipe Products
Click to view available options
:
Dry wiper; 12 x 12 in.
Description
- Double-knit, no-run interlock construction of 100% continuous-filament polyester
- Two-ply, sealed-border wiper with highest sorbency
- Ideal wiper for use in multiple environments
- Excellent for picking up large amounts of liquids and for applying chemicals
- Abrasion-resistant wiper with soft, flexible hand
- Lot-to-lot traceability
Order Info
Available dry or sterile. Basis weight: 185g/m2.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction