missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tft.-45.6 Fixed Angle Rotor

Product Code. 12151640 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
40 x 6 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12151640 40 x 6 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12151640 Supplier Thermo Scientific™ Supplier No. 11637
  • ONLINE EXCLUSIVE PRICE
    27540.00 €
    27000.00 € / Pack of 1
    Save 540.00 €
    1% Off
 Restricted Item
Estimated Shipment: 10-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Conduct a variety of applications using the versatile Thermo Scientific™ TFT-45.6 Fixed Angle Rotor, which includes a range of capabilities.

Compatible with:

Thermo Scientific Sorvall WX Ultra Series Floor Model Centrifuge

Recommended for:

Serum lipoprotein flotation, multiplesample DNA and virus isolation.

TRUSTED_SUSTAINABILITY

Specifications

Description TFT-45.6 Fixed Angle Rotor
Material Titanium
Type Fixed Angle Rotor
Capacity 40 x 6 mL
Max. RCF 238,859 x g (Outer Row), 204,252 x g (Inner Row)
Max. Speed 45,000 rpm
Radius (Metric) 105.6 mm Outer, 90.3 mm Inner
Radius (Metric) Min. 76.0 mm Outer, 60.7 mm Inner
K Factor 44.1 Outer Row/49.6 Inner Row
Max. Tube Length 66 mm
Max. Tube Diameter 13 mm
Net Weight 24.3 lb. (10.9 kg)
For Use With (Equipment) Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle 20°
Avg. RCF 205,383 x g Outer/170,755 x g Inner
Min. RCF 171,906 x g Outer; 137,299 x g Inner
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.