missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TGX 221

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TGX 221 is a potent and selective PI 3-kinase β inhibitor (IC50 values are 0.007, 0.1, 3.5 and 5 μM for the β, δ, γ and α isoforms, respectively). Shows >1,000-fold selectively for PI 3-kinase β over a range of other kinases. Inhibits thrombus formation in animal models.
Specifications
Specifications
| Chemical Name or Material | 7-Methyl-2-(4-morpholinyl)-9-[1-(phenylamino)ethyl]-4H-pyrido[1,2-a]pyrimidin-4-one |
| CAS | 663619-89-4 |
| Quantity | 10 mg |
| Target | PI 3-Kinase Inhibitors |
| Molecular Formula | C21H24N4O2 |
| Formula Weight | 364.44 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction