missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TH 10785
GREENER_CHOICE

Product Code. 18797406
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18797406 50 mg 50mg
18746956 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18797406 Supplier Tocris Bioscience™ Supplier No. 7778/50
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

OGG1 activator

TH 10785 is an 8-oxo guanine DNA glycosylase 1 (OGG1) activator. It binds in the active site of OGG1 and increases its recruitment to oxidative DNA damage and its repair.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Cyclohexyl-2-cyclopropyl-4-quinazolinamine
CAS 1002801-51-5
Quantity 50 mg
Target Glycosylase Activators
Molecular Formula C17H21N3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.