missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Thalidomide

Product Code. 18702654
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18702654 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702654 Supplier R&D Systems™ Supplier No. 0652/100
  • ONLINE EXCLUSIVE PRICE
    147.32 €
    132.30 € / 100mg
    Save 15.02 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Binds cereblon; also TNF-α synthesis inhibitor

Thalidomide is a selective inhibitor of tumor necrosis factor α (TNF-α) synthesis. Anti-inflammatory; promotes T-cell activation. Predicted to reduce cytokine release in SARS-CoV-2 infection. Also binds cereblon, inhibiting ubiquitin ligase activity. Teratogenic and sedative-hypnotic. Promotes degradation of transcription factor SALL4.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-35-1
Quantity 100 mg
Product Line Tocris
Molecular Formula N-(2,6-dioxo-3-piperidinyl)phthalimide
Product Type Thalidomide
Purity 99%
Molecular Weight (g/mol) M.W. (Observed): 258.23
Inhibitors Thalidomide

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.