missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Thalidomide, 99+%

Product Code. 15454669
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454669 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15454669 Supplier Thermo Scientific Alfa Aesar Supplier No. J62266.MC
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50-35-1 | C13H10N2O4 | 258.23 g/mol

Teratogenic compound found to inhibit HIV-1 replication and FGF-induced angiogenesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 50-35-1
Color White
Molecular Formula C13H10N2O4
MDL Number MFCD00153873
Quantity 100 mg
UN Number UN2811
Merck Index 14,9255
Synonym thalidomide, thalomid, +/--thalidomide, sedoval, 2-2,6-dioxopiperidin-3-yl isoindoline-1,3-dione, corronarobetin, psycholiquid, psychotablets, theophilcholine, algosediv
Solubility Information Soluble in DMSO; Insoluble in water or ethanol
InChI Key UEJJHQNACJXSKW-UHFFFAOYNA-N
SMILES O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12
IUPAC Name 2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Weight (g/mol) 258.23
PubChem CID 5426
ChEBI CHEBI:74947
Formula Weight 258.23
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material (+/-)-Thalidomide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.