missing translation for 'onlineSavingsMsg'
Learn More

Thallic trifluoroacetate, tech., Thermo Scientific Chemicals

Product Code. p-3777091 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10157990

Brand: Thermo Scientific Acros 420760250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23586-53-0
Molecular Formula C6F9O6Tl
Molecular Weight (g/mol) 543.42
MDL Number MFCD00000414
InChI Key PSHNNUKOUQCMSG-UHFFFAOYSA-K
Synonym thallium iii trifluoroacetate, acetic acid, trifluoro-, thallium 3+ salt, acetic acid,2,2,2-trifluoro-, thallium 3+ salt 3:1, thallium 3+ ; 2,2,2-trifluoroacetate, thallium tris trifluoroacetate, trifluoroacetic acid thallium iii, thallic trifluoroacetate, tech, thallium 3+ tritrifluoroacetate
PubChem CID 90200
IUPAC Name thallium(3+);2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Tl+3]

Specifications

Color Beige
Physical Form Fine Powder
Quantity 25 g
Linear Formula (CF3COO)3Tl
Beilstein 02, II, 186
Fieser 03,286; 04,498; 05,658; 06,579; 07,365; 08,478; 10,397; 05,654
Formula Weight 543.42
Grade Technical
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Thallic trifluoroacetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thallic trifluoroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.