missing translation for 'onlineSavingsMsg'
Learn More

Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water

Product Code. 11326498
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326498 25 g 25g
11336498 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11326498 Supplier Thermo Scientific Alfa Aesar Supplier No. 012131.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-28-0 | Tl | 204.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in water
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]

Specifications

Color Blue-White
Quantity 25 g
Odor Odorless
Particle Size 6mm (0.2 in.) & down
UN Number UN1707
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Thallium granules

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.