missing translation for 'onlineSavingsMsg'
Learn More

Thallium Sulfate, TRC

Product Code. 30456188
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456188 500 mg 500mg
30450032 5 g -
30448993 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456188 Supplier Toronto Research Chemicals Supplier No. TRCT790543500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4 S . 2 Tl
Molecular Weight (g/mol) 504.83
Synonym Sulfuric acid, thallium(1+) salt (1:2) (ACI), Sulfuric acid, dithallium(1+) salt (8CI, 9CI), Thallium sulfate (Tl2(SO4)) (6CI), C.F.S., CSF-Giftweizen, Dithallium sulfate, Dithallium(1+) sulfate, Dithallium(I) sulfate, M 7-Giftkoerner, Rattengiftkonserve, Thallium sulfate, Thallium(I) sulfate, Thallous sulfate, Zelio
IUPAC Name thallium(1+);sulfate
SMILES [Tl+].[Tl+].[O-]S(=O)(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/H2O4S.2Tl/c1-5(2,3)4;;/h(H2,1,2,3,4);;/q;2*+1/p-2
Formula Weight 505.9006
Chemical Name or Material Thallium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.