missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Thapsigargin

Product Code. 18722754
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722754 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722754 Supplier R&D Systems™ Supplier No. 1138/1
  • ONLINE EXCLUSIVE PRICE
    160.00 €
    151.20 € / 1mg
    Save 8.80 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent inhibitor of SERCA ATPase

Potent inhibitor of sarco-endoplasmic reticulum Ca2+-ATPases. Causes ER stress; can be used to induce autophagy in mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67526-95-8
Quantity 1 mg
Product Line Tocris
Molecular Formula C34H50O12
Formula Weight 650.76
Purity >97%
Molecular Weight (g/mol) 650.76
Chemical Name or Material Thapsigargin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.