missing translation for 'onlineSavingsMsg'
Learn More

Thapsigargin, MedChemExpress

Product Code. 16448377
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448377 5 mg 5mg
16411349 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448377 Supplier Medchem Express Supplier No. HY13433
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thapsigargin, an endoplasmic reticulum (ER) stress inducer, is an inhibitor of microsomal Ca2+-ATPase. Thapsigargin efficiently inhibits coronavirus (HCoV-229E, MERS-CoV, SARS-CoV-2) replication in different cell types.

Thapsigargin, an endoplasmic reticulum (ER) stress inducer, is an inhibitor of microsomal Ca2+-ATPase. Thapsigargin efficiently inhibits coronavirus (HCoV-229E, MERS-CoV, SARS-CoV-2) replication in different cell types.

Specifications

Chemical Name or Material Thapsigargin
CAS 67526-95-8
Color White
Purity Grade Notes Research
Molecular Formula C34H50O12
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (153.67 mM; Need ultrasonic)
SMILES CCCCCCCC(O[C@@H]1[C@@H](OC(/C(C)=C\C)=O)C(C)=C2[C@@]1([H])[C@@](C)(OC(C)=O)C[C@H](OC(CCC)=O)[C@@]([C@@]3(O)C)(O)[C@@]2([H])OC3=O)=O
Molecular Weight (g/mol) 650.75
Formula Weight 650.75
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.