Learn More
STEMCELL Technologies The Big Easy' EasySep™ Magnet
Product Code. 16604402
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
Each
Product Code. Quantity Unit Size
16604402 1 x 109 Cells Each
1 options
Product Code. 16604402 Supplier STEMCELL Technologies Supplier No. 18001
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Immunomagnetic column-free magnet
  • The Big Easy EasySep™ Magnet is designed for cell separation procedures using EasySep™ reagents when processing a large number of cells, or when using RoboSep™
  • The Big Easy EasySep™ Magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ Magnetic Particles without the use of columns
  • The Big Easy EasySep™ Magnet is designed to hold a standard 14 mL (17 x 95 mm) polystyrene tube
Specifications
For Use With (Application) Cell Isolation
Quantity 1 x 109 Cells