Learn More
Abeba™ The Original 7081 Shoes
Product Code. 15692508
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
Unit Size:
1 pair
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15692508 38 1 pair
15662508 35 1 pair
15672508 36 1 pair
15682508 37 1 pair
15602518 39 1 pair
15612518 40 1 pair
15622518 41 1 pair
15632518 42 1 pair
15642518 43 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15692508 Supplier Abeba™ Supplier No. 7081/38
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Animal fats, oil and fuel resistant. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather
  • Light blue/White
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB FO SRC
Color Light Blue, White
Material Leather
Size 38
Sole Features Slip-resistant
Style 7081
Resistant To Animal fats, oil and fuel