missing translation for 'onlineSavingsMsg'
Learn More

Theobromine, 99%

Product Code. 10175213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10175213 1 kg Plastic bottle 1kg
10124920 100 g Plastic bottle 100g
10033310 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10175213 Supplier Thermo Scientific Acros Supplier No. 258820010
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Theobromine, CAS # 83-67-0, is a methylxanthine alkaloid that is present in large quantities in cocoa and is a derivative of caffein. | CAS: 83-67-0 | C7H8N4O2 | 180.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Theobromine is a methylxanthine alkaloid that is a derivative of caffeine

Applications
• Theobromine is a phosphodiesterase inhibitor, weak adenosine receptor antagonist, and smooth muscle relaxant
• As a catabolic product of caffeine, it has antioxidant and prooxidant properties
• It can be used as a standard for quantification of theobromine

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-67-0
Molecular Formula C7H8N4O2
Molecular Weight (g/mol) 180.17
MDL Number MFCD00022830
InChI Key YAPQBXQYLJRXSA-UHFFFAOYSA-N
Synonym theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
PubChem CID 5429
ChEBI CHEBI:28946
IUPAC Name 3,7-dimethylpurine-2,6-dione
SMILES CN1C=NC2=C1C(=O)NC(=O)N2C

Specifications

Melting Point 290.0°C to 295.0°C
Color White
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 1 kg
Beilstein 26, 2336
Merck Index 15, 9433
Solubility Information Solubility in water: slightly soluble. Other solubilities: moderately soluble in ammonia, practically insoluble in benzene, ether, carbon, tetrachloride and chloroform
Sublimation Point 290.0 °C
Formula Weight 180.17
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material Theobromine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.