missing translation for 'onlineSavingsMsg'
Learn More

Theobromine, TRC

Product Code. 30007751
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007751 5 g 5g
17981995 1 g 1g
17951975 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007751 Supplier Toronto Research Chemicals Supplier No. TRCT3438005G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-67-0
Molecular Formula C7 H8 N4 O2
Molecular Weight (g/mol) 180.16
Synonym Theobromine, Theophylline-Ethylenediamine Imp. G (EP), Pentoxifylline Imp. A (EP), Caffeine Imp. D (EP)
IUPAC Name 3,7-dimethylpurine-2,6-dione
SMILES CN1C(=O)NC(=O)c2c1ncn2C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)
Formula Weight 180.0647
Percent Purity >95%
Chemical Name or Material Theobromine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.