missing translation for 'onlineSavingsMsg'
Learn More

Theophylline-7-acetic acid, 98%, Thermo Scientific Chemicals

Product Code. 15512363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15512363 25 g 25g
15522363 100 g 100g
15532363 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15512363 Supplier Thermo Scientific Acros Supplier No. 459250250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-37-9
Molecular Formula C9H9N4NaO4
Molecular Weight (g/mol) 260.19
MDL Number MFCD00022832
InChI Key MSFVZSOKOXZSME-UHFFFAOYSA-M
Synonym acefylline, theophylline-7-acetic acid, acephylline, theophyllineacetic acid, carboxymethyltheophylline, 7-carboxymethyl theophylline, 7-theophyllinylacetic acid, theophyllin-7-ylacetic acid, 7-theophyllinessigsaeure, 7-theophyllineacetic acid
PubChem CID 69550
IUPAC Name 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetic acid
SMILES [Na+].CN1C2=C(N(CC([O-])=O)C=N2)C(=O)N(C)C1=O

Specifications

Melting Point 270.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 238.2
Percent Purity 98%
Chemical Name or Material Theophylline-7-acetic acid
compliance-icons
Product Identifier
  • Theophyllin-7-ylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.